Assay Details
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017348.2 | NP_059044.1 | 10 - 11 | 2084 | 88 | |
XM_006229611.1 | XP_006229673.1 | 9 - 10 | 1981 | 88 | ||
GenBank mRNA | BC168238.1 | - | 10 - 11 | 2062 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
