Assay Details
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017348.2 | NP_059044.1 | 3 - 4 | 1232 | 77 | |
XM_006229611.1 | XP_006229673.1 | 3 - 4 | 1264 | 77 | ||
GenBank mRNA | BC168238.1 | - | 3 - 4 | 1210 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
