Assay Details
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022259.1 | NP_071595.1 | 2 - 3 | 440 | 98 | |
XM_006250259.2 | XP_006250321.1 | 2 - 3 | 1819 | 98 | ||
GenBank mRNA | AF156988.1 | - | 2 - 3 | 500 | 98 | |
AF156989.1 | - | 2 - 3 | 440 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
