Assay Details
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022259.1 | NP_071595.1 | 4 - 5 | 826 | 94 | |
XM_006250259.2 | XP_006250321.1 | 4 - 5 | 2205 | 94 | ||
GenBank mRNA | AF156988.1 | - | 4 - 5 | 755 | 94 | |
AF156989.1 | - | 4 - 5 | 826 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
