Assay Details
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022259.1 | NP_071595.1 | 5 - 6 | 973 | 75 | |
GenBank mRNA | AF156988.1 | - | 5 - 6 | 902 | 75 | |
AF156989.1 | - | 5 - 6 | 973 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
