Assay Details
Gene Symbol: Rhoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057132.3 | NP_476473.1 | 2 - 3 | 274 | 100 | |
XM_006243699.1 | XP_006243761.1 | 4 - 5 | 542 | 100 | ||
XM_006243700.1 | XP_006243762.1 | 4 - 5 | 517 | 100 | ||
XM_006243701.1 | XP_006243763.1 | 4 - 5 | 637 | 100 | ||
GenBank mRNA | AY026068.1 | - | 2 - 3 | 274 | 100 | |
AY026069.1 | - | 2 - 3 | 274 | 100 | ||
BC061732.1 | - | 4 - 5 | 451 | 5598621 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
