Assay Details
Gene Symbol: Rhoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057132.3 | NP_476473.1 | 3 - 4 | 407 | 82 | |
XM_006243699.1 | XP_006243761.1 | 5 - 6 | 675 | 82 | ||
XM_006243700.1 | XP_006243762.1 | 5 - 6 | 650 | 82 | ||
XM_006243701.1 | XP_006243763.1 | 5 - 6 | 770 | 82 | ||
GenBank mRNA | AY026068.1 | - | 3 - 4 | 407 | 82 | |
AY026069.1 | - | 3 - 4 | 407 | 82 | ||
BC061732.1 | - | 5 - 6 | 584 | 5598621 | 82 | |
D84477.1 | - | 3 - 4 | 377 | 82 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
