Assay Details
Gene Symbol: LOC689600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106351.1 | NP_001099821.1 | 1 - 2 | 160 | 104 |
Gene Symbol: RGD1564599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173448.1 | NP_001166919.1 | 1 - 2 | 160 | 104 | |
XM_006229537.2 | XP_006229599.1 | 1 - 2 | 166 | 104 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
