Assay Details
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039036.1 | NP_001034125.1 | 1 | 53 | 78 | |
GenBank mRNA | BC098859.1 | - | 1 | 53 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC098859.1, NM_001039036.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
