Assay Details
Gene Symbol: Rps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031108.3 | NP_112370.2 | 1 | 45 | 97 | |
GenBank mRNA | BC060560.1 | - | 1 | 50 | 97 |
Gene Symbol: LOC103689992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758836.1 | XP_008757058.1 | 75 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC060560.1, NM_031108.3, XM_008758836.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
