Assay Details
Gene Symbol: Mir148b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031782.1 | 1 | 47 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
coatomer protein complex, subunit zeta 1 |
Copz1 | Sense | Chr. 7 - 144900091 - 144926299 on NCBI Build Rnor_6.0 | ||
Downstream | G protein-coupled receptor 84 | Gpr84 | 22.5kb | Anti-sense | Chr. 7 - 144934606 - 144936865 on NCBI Build Rnor_6.0 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
