Assay Details
Gene Symbol: Tyw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033170.1 | 2 - 3 | 480 | 85 | ||
GenBank mRNA | BC158619.1 | - | 2 - 3 | 480 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | similar to CG31122-PA | RGD1306941 | -11.8kb | Anti-sense | Chr. 9 - 64066579 - 64075197 on NCBI Build Rnor_6.0 | |
Downstream | similar to hypothetical protein FLJ22555 | MGC94335 | 4.8kb | Anti-sense | Chr. 9 - 64095978 - 64103908 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
