Assay Details
Gene Symbol: Ptpn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177593.1 | NP_001171064.1 | 8 - 9 | 1108 | 78 | |
NM_013088.2 | NP_037220.2 | 8 - 9 | 1108 | 78 | ||
GenBank mRNA | D83016.1 | - | 2 - 3 | 157 | 78 | |
U05963.1 | - | 8 - 9 | 1039 | 78 | ||
U09307.1 | - | 8 - 9 | 1033 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
