Assay Details
Gene Symbol: Mettl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191814.1 | NP_001178743.1 | 6 - 7 | 1180 | 90 | |
XM_006245658.2 | XP_006245720.1 | 7 - 8 | 1838 | 90 | ||
XM_006245661.2 | XP_006245723.1 | 6 - 7 | 1776 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
