Assay Details
Gene Symbol: C1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191891.1 | NP_001178820.1 | 1 - 2 | 163 | 53 | |
XM_006252134.2 | XP_006252196.1 | 2 - 3 | 943 | 53 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
