Assay Details
Gene Symbol: Synpo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191963.1 | NP_001178892.1 | 2 - 3 | 383 | 63 | |
XM_008761470.1 | XP_008759692.1 | 3 - 4 | 390 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
