Assay Details
Gene Symbol: Gfral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191998.1 | NP_001178927.1 | 8 - 9 | 1114 | 91 | |
XM_006243416.2 | XP_006243478.1 | 9 - 10 | 3154 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
