Assay Details
Gene Symbol: Tmem80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045202.1 | 4 - 5 | 973 | 88 | ||
GenBank mRNA | CB546081.1 | - | 4 - 5 | 320 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DEAF1 transcription factor | Deaf1 | -5.5kb | Anti-sense | Chr. 1 - 214283787 - 214317466 on NCBI Build Rnor_6.0 | |
Downstream | EPS8-like 2 | Eps8l2 | 1.7kb | Sense | Chr. 1 - 214327992 - 214353466 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
