Assay Details
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108598.2 | NP_001102068.2 | 6 - 7 | 492 | 74 | |
GenBank mRNA | CB576376.1 | - | 6 - 7 | 369 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
