Assay Details
Gene Symbol: Nfatc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244933.1 | NP_001231862.1 | 8 - 9 | 2398 | 66 | |
GenBank mRNA | FQ222003.1 | - | 8 - 9 | 2398 | 66 | |
FQ223020.1 | - | 8 - 9 | 2398 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
