Assay Details
Gene Symbol: Smndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_051995.1 | 3 - 4 | 452 | 92 | ||
GenBank mRNA | BC098697.1 | - | 3 - 4 | 414 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MAX interactor 1 | Mxi1 | MXI-WR | -10.0kb | Anti-sense | Chr. 1 - 274030748 - 274090071 on NCBI Build Rnor_6.0 |
Downstream | dual specificity phosphatase 5 | Dusp5 | Cpg21 | 144.2kb | Anti-sense | Chr. 1 - 274245184 - 274258595 on NCBI Build Rnor_6.0 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
