Assay Details
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014245.2 | NP_001014267.1 | 3 - 4 | 440 | 86 | |
XM_006231061.2 | XP_006231123.1 | 5 - 6 | 595 | 86 | ||
XM_006231062.2 | XP_006231124.1 | 4 - 5 | 583 | 86 | ||
XM_008760238.1 | XP_008758460.1 | 4 - 5 | 514 | 86 | ||
XM_008760239.1 | XP_008758461.1 | 4 - 5 | 514 | 86 | ||
XM_008760240.1 | XP_008758462.1 | 3 - 4 | 401 | 86 | ||
GenBank mRNA | BC083864.1 | - | 3 - 4 | 443 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
