Assay Details
Gene Symbol: Cpsf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014245.2 | NP_001014267.1 | 6 - 7 | 1113 | 62 | |
XM_006231061.2 | XP_006231123.1 | 8 - 9 | 1295 | 62 | ||
XM_006231062.2 | XP_006231124.1 | 7 - 8 | 1283 | 62 | ||
XM_008760238.1 | XP_008758460.1 | 7 - 8 | 1214 | 62 | ||
XM_008760239.1 | XP_008758461.1 | 7 - 8 | 1187 | 62 | ||
XM_008760240.1 | XP_008758462.1 | 6 - 7 | 1101 | 62 | ||
GenBank mRNA | BC083864.1 | - | 6 - 7 | 1116 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
