Assay Details
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076553.2 | NP_001070021.1 | 5 - 6 | 556 | 62 | |
NM_001271158.1 | NP_001258087.1 | 4 - 5 | 391 | 62 | ||
NM_012627.3 | NP_036759.2 | 4 - 5 | 451 | 62 | ||
XM_006256511.2 | XP_006256573.1 | 5 - 6 | 1175 | 62 | ||
XM_006256512.2 | XP_006256574.1 | 6 - 7 | 678 | 62 | ||
XM_006256513.2 | XP_006256575.1 | 5 - 6 | 533 | 62 | ||
XM_006256515.2 | XP_006256577.1 | 5 - 6 | 1098 | 62 | ||
XM_008772976.1 | XP_008771198.1 | 6 - 7 | 815 | 62 | ||
XM_008772977.1 | XP_008771199.1 | 6 - 7 | 1528 | 62 | ||
XM_008772978.1 | XP_008771200.1 | 5 - 6 | 1265 | 62 | ||
GenBank mRNA | AF413572.1 | - | 5 - 6 | 539 | 62 | |
BC078880.1 | - | 4 - 5 | 391 | 7111554 | 62 | |
FQ231722.1 | - | 4 - 5 | 447 | 62 | ||
M64092.1 | - | 4 - 5 | 408 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
