Assay Details
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270688.1 | NP_001257617.1 | 4 - 5 | 234 | 83 | |
NM_001270689.1 | NP_001257618.1 | 4 - 5 | 262 | 83 | ||
NM_021909.2 | NP_068709.2 | 4 - 5 | 338 | 83 | ||
GenBank mRNA | BC060571.1 | - | 4 - 5 | 226 | 83 | |
FM122563.1 | - | 4 - 5 | 212 | 83 | ||
FQ220058.1 | - | 4 - 5 | 262 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
