Assay Details
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270838.1 | NP_001257767.1 | 8 - 9 | 1110 | 70 | |
NM_001270840.1 | NP_001257769.1 | 8 - 9 | 1107 | 70 | ||
GenBank mRNA | AB204897.1 | - | 7 - 8 | 958 | 70 | |
AB204898.1 | - | 7 - 8 | 961 | 70 | ||
BC089763.1 | - | 8 - 9 | 1107 | 7324318 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
