Assay Details
Gene Symbol: Hnf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270931.1 | NP_001257860.1 | 6 - 7 | 832 | 58 | |
NM_001270933.1 | NP_001257862.1 | 6 - 7 | 832 | 58 | ||
NM_022180.2 | NP_071516.2 | 5 - 6 | 792 | 58 | ||
XM_006235515.2 | XP_006235577.1 | 5 - 6 | 787 | 58 | ||
XM_006235516.2 | XP_006235578.1 | 5 - 6 | 787 | 58 | ||
GenBank mRNA | CK483260.1 | - | 5 - 6 | 621 | 58 | |
D10554.1 | - | 5 - 6 | 668 | 58 | ||
EF193390.1 | - | 5 - 6 | 613 | 58 | ||
EF193392.1 | - | 5 - 6 | 594 | 58 | ||
FJ608816.1 | - | 6 - 7 | 832 | 58 | ||
FJ608817.1 | - | 6 - 7 | 832 | 58 | ||
FJ608818.1 | - | 6 - 7 | 832 | 58 | ||
X57133.1 | - | 5 - 6 | 683 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
