Assay Details
Gene Symbol: Hnf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270931.1 | NP_001257860.1 | 7 - 8 | 912 | 59 | |
NM_001270933.1 | NP_001257862.1 | 7 - 8 | 912 | 59 | ||
NM_022180.2 | NP_071516.2 | 6 - 7 | 872 | 59 | ||
XM_006235515.2 | XP_006235577.1 | 6 - 7 | 867 | 59 | ||
XM_006235516.2 | XP_006235578.1 | 6 - 7 | 867 | 59 | ||
GenBank mRNA | D10554.1 | - | 6 - 7 | 748 | 59 | |
EF193390.1 | - | 6 - 7 | 693 | 59 | ||
EF193392.1 | - | 6 - 7 | 674 | 59 | ||
FJ608816.1 | - | 7 - 8 | 912 | 59 | ||
FJ608817.1 | - | 7 - 8 | 912 | 59 | ||
FJ608818.1 | - | 7 - 8 | 912 | 59 | ||
X57133.1 | - | 6 - 7 | 763 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
