Assay Details
Gene Symbol: Cnnm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271050.1 | NP_001257979.1 | 4 - 5 | 1846 | 66 | |
XM_006244827.2 | XP_006244889.1 | 4 - 5 | 1792 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
