Assay Details
Gene Symbol: Tnfrsf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271235.1 | NP_001258164.1 | 6 - 7 | 640 | 68 | |
XM_008769487.1 | XP_008767709.1 | 6 - 7 | 618 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
