Assay Details
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271330.1 | NP_001258259.1 | 2 - 3 | 446 | 60 | |
GenBank mRNA | FM045749.1 | - | 2 - 3 | 451 | 60 | |
FM067602.1 | - | 2 - 3 | 212 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
