Assay Details
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271330.1 | NP_001258259.1 | 5 - 6 | 759 | 69 | |
GenBank mRNA | FM067602.1 | - | 5 - 6 | 525 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
