Assay Details
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271330.1 | NP_001258259.1 | 9 - 10 | 1205 | 58 | |
GenBank mRNA | FM093163.1 | - | 2 - 3 | 163 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
