Assay Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012678.2 | NP_036810.1 | 3 - 4 | 411 | 108 | |
GenBank mRNA | FQ225054.1 | - | 422 | 108 | ||
FQ225082.1 | - | 422 | 108 | |||
J02780.1 | - | 3 - 4 | 414 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
