Assay Details
Gene Symbol: Tpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012678.2 | NP_036810.1 | 5 - 6 | 560 | 94 | |
GenBank mRNA | FQ225054.1 | - | 571 | 94 | ||
FQ225082.1 | - | 571 | 94 | |||
J02780.1 | - | 5 - 6 | 563 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
