Assay Details
Gene Symbol: Atf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012912.2 | NP_037044.1 | 2 - 3 | 395 | 95 | |
GenBank mRNA | BC078903.1 | - | 2 - 3 | 395 | 95 | |
M63282.1 | - | 2 - 3 | 400 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
