Assay Details
Gene Symbol: Acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013084.2 | NP_037216.1 | 8 - 9 | 1005 | 63 | |
XM_008759865.1 | XP_008758087.1 | 8 - 9 | 1090 | 63 | ||
XM_008759866.1 | XP_008758088.1 | 8 - 9 | 1090 | 63 | ||
GenBank mRNA | U64451.1 | - | 8 - 9 | 1006 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
