Assay Details
Gene Symbol: Prkar2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019264.2 | NP_062137.1 | 5 - 6 | 643 | 95 | |
XM_006243712.2 | XP_006243774.1 | 6 - 7 | 828 | 95 | ||
XM_006243713.2 | XP_006243775.1 | 6 - 7 | 642 | 95 | ||
XM_008766478.1 | XP_008764700.1 | 5 - 6 | 593 | 95 | ||
GenBank mRNA | AF533978.1 | - | 5 - 6 | 647 | 95 | |
J02934.1 | - | 5 - 6 | 441 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
