Assay Details
Gene Symbol: Wnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053402.2 | NP_445854.1 | 2 - 3 | 315 | 69 | |
GenBank mRNA | AF188608.1 | - | 2 - 3 | 316 | 69 | |
BC098752.1 | - | 2 - 3 | 336 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
