Assay Details
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053407.3 | NP_445859.2 | 13 - 14 | 1131 | 114 | |
GenBank mRNA | BC061540.1 | - | 13 - 14 | 1134 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P113699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149359 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P113699 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P113699 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P113699 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12798 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P113699 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69175 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84958 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73027 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10788355 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788359 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788353 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788358 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788354 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788351 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788352 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788360 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788350 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788361 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788363 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788362 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788364 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788356 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788357 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10788348 | - | - |
Rat Genome 230 2.0 Array | 1374770_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374770_at | - | - |
Rat Genome 230 2.0 Array | 1369077_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369077_at | - | - |
Affymetrix HT Rat Focus Array | 1369077_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374770_at | - | - |
Affymetrix HT Rat Focus Array | 1369077_at | - | - |
Affymetrix HT Rat Focus Array | 1374770_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600004949.rn.1 | TC1600000621.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600004953.rn.1 | TC1600000621.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600004951.rn.1 | TC1600000621.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600004952.rn.1 | TC1600000621.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1600004950.rn.1 | TC1600000621.rn.1 | RTA TC |
Rat Expression Set 230 | 1374770_at | - | - |
Rat Expression Set 230 | 1369077_at | - | - |
Rat Expression Set 230 | 1374770_PM_at | - | - |
Rat Expression Set 230 | 1369077_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788348 | - | - |
Rat Genome U34 Set | rc_AA800062_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21562817 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368380 | - | - |
RatRef-12 Expression BeadChip | ILMN_48736 | - | - |
