Assay Details
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053407.3 | NP_445859.2 | 5 - 6 | 408 | 117 | |
GenBank mRNA | AF214647.1 | - | 5 - 6 | 388 | 117 | |
BC061540.1 | - | 5 - 6 | 411 | 6919949 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
