Assay Details
Gene Symbol: Timm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_075101.1 | 2 - 3 | 216 | 117 | ||
GenBank mRNA | DV726219.1 | - | 2 - 3 | 186 | 117 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | translocase of outer mitochondrial membrane 20 homolog (yeast)-like | Tomm20l | Anti-sense | Chr. 6 - 93538827 - 93549533 on NCBI Build Rnor_6.0 | ||
Downstream | similar to Protein KIAA0586 | LOC690035 | 5.3kb | Anti-sense | Chr. 6 - 93562778 - 93667857 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
