Assay Details
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277228.1 | NP_001264157.1 | 2 - 3 | 658 | 62 | |
GenBank mRNA | BC085943.1 | - | 2 - 3 | 658 | 62 | |
BC168987.1 | - | 2 - 3 | 620 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
