Assay Details
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277228.1 | NP_001264157.1 | 5 - 6 | 1114 | 75 | |
GenBank mRNA | BC168987.1 | - | 5 - 6 | 1076 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
