Assay Details
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080896.1 | NP_543172.1 | 9 - 10 | 1046 | 68 | |
GenBank mRNA | AF367468.1 | - | 9 - 10 | 1046 | 68 | |
FQ212957.1 | - | 6 - 7 | 625 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
