Assay Details
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751297.3 | XP_003751345.1 | 7 - 8 | 1628 | 89 | |
XM_008769895.1 | XP_008768117.1 | 6 - 7 | 2611 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
