Assay Details
Gene Symbol: Faf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017445.1 | NP_001017445.1 | 5 - 6 | 378 | 74 | |
XM_006253617.2 | XP_006253679.1 | 4 - 5 | 403 | 74 | ||
GenBank mRNA | BC079356.1 | - | 4 - 5 | 377 | 74 | |
BC091224.1 | - | 5 - 6 | 378 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
