Assay Details
Gene Symbol: Mir28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031834.1 | 1 | 44 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tumor protein p63 regulated 1 | Tprg1 |
RGD1306226, Svap30, Tprg |
-518.8kb | Sense | Chr. 11 - 78733046 - 78861745 on NCBI Build Rnor_6.0 |
Downstream | B-cell CLL/lymphoma 6 | Bcl6 | 875.1kb | Anti-sense | Chr. 11 - 80255722 - 80279077 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
