Assay Details
Gene Symbol: Gtf2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022208.1 | NP_071544.1 | 8 - 9 | 1017 | 95 | |
XM_006240410.2 | XP_006240472.1 | 8 - 9 | 1292 | 95 | ||
GenBank mRNA | AF000943.1 | - | 8 - 9 | 1017 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
