Assay Details
Gene Symbol: Poli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106137.1 | NP_001099607.1 | 9 | 1369 | 73 | |
XM_006254900.2 | XP_006254962.1 | 9 | 1290 | 73 | ||
XM_006254901.2 | XP_006254963.1 | 8 | 1232 | 73 | ||
XM_008772110.1 | XP_008770332.1 | 6 | 1224 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
